missing translation for 'onlineSavingsMsg'
Learn More

Varicella zoster virus gE Mouse anti-Virus, Clone: VA030, Abnova™

Product Code. 16032186
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16032186 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16032186 Supplier Abnova Supplier No. MAB13662.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against varicella zoster virus (VZV) gE.

  • High specificity
  • High reactivity

Specifications

Antigen Varicella zoster virus gE
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone VA030
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against varicella zoster virus (VZV) gE.
Dilution ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4.
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with gE of varicella zoster virus (VZV).
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.