missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97417F
This item is not returnable.
View return policy
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative).
Specifications
| Vanilloid R1/TRPV1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative) | |
| Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Invertebrate, Primate | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TRPV1 | |
| A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] | |
| 0.1 mL | |
| Neuroscience, Phospho Specific, Sensory Systems | |
| 7442 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction