missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanilloid R-like 3/TRPV3 Antibody (S15/4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-12909-0.025mg
This item is not returnable.
View return policy
Description
Vanilloid R-like 3/TRPV3 Monoclonal specifically detects Vanilloid R-like 3/TRPV3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| Vanilloid R-like 3/TRPV3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000 | |
| TRPV3 | |
| Synthetic peptide amino acids 774-791 (C-terminus) of rat TrpV3 | |
| 0.025 mg | |
| Primary | |
| Detects approx 70 kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| S15/4 | |
| Unconjugated | |
| transient receptor potential cation channel subfamily V member 3, transient receptor potential cation channel, subfamily V, member 3, TrpV3, vanilloid receptor 3, Vanilloid receptor-like 3, vanilloid receptor-related osmotically activated channel protein, VRL3, VRL-3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 162514 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction