Learn More
Vanillin Melting Point Standard, Reagecon™
BIOPHARMA
Catalog Number 13552100
Click to view available options
Quantity:
1 g
Packaging:
1g
Packaging 1g
Quantity 1 g
compliance-icons
Product Identifier
  • MELTING POINT STANDARD VANILLIN 81 to 83°C
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.