missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VAMP-2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30983-250ug
This item is not returnable.
View return policy
Description
VAMP-2 Polyclonal antibody specifically detects VAMP-2 in Human,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| VAMP-2 | |
| Polyclonal | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2) | |
| Full length VAMP-2 | |
| 250 μg | |
| Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6844 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction