missing translation for 'onlineSavingsMsg'
Learn More

(+)-Valencene 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30326525
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30326525

Brand: Dr. Ehrenstorfer DREGA09010079IP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valencene
CAS 7-3-4630
Analyte or Component Names Valencene
Shipping Condition +20°C
Molecular Formula C15 H24
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym (1R,7R,8aS)-1,2,3,5,6,7,8,8a-Octahydro-1,8a-dimethyl-7-(1-methylethenyl)naphthalene, 4βH,5α-Eremophila-1(10),11-diene, [1R-(1α,7β,8aα)]-1,2,3,5,6,7,8,8a-Octahydro-1,8a-dimethyl-7-(1-methylethenyl)naphthalene, Valencene, (+)-Valencene, 4βH,5α-Eremophila-1(10),11-diene, NSC 148969, Valencen
InChI Formula InChI=1S/C15H24/c1-11(2)13-8-9-14-7-5-6-12(3)15(14,4)10-13/h7,12-13H,1,5-6,8-10H2,2-4H3/t12-,13-,15+/m1/s1
SMILES C[C@@H]1CCC=C2CC[C@H](C[C@@]12C)C(=C)C
IUPAC Name (3R,4aS,5R)-4a,5-dimethyl-3-prop-1-en-2-yl-2,3,4,5,6,7-hexahydro-1H-naphthalene
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 204.35
Quantity 1 mL
Formula Weight 204.1878 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.