Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Product Code. p-4522205 Shop All Thermo Scientific Products
Click to view available options
Description:
Ferrule
Male Nut
Two-piece Fingerlok Nut with PEEK Ferrule
Valco 10-32 male hex nut
Quantity:
1 Each
10 Pack
Unit Size:
Each
Pack of 10
Product Code. 10639865
Brand: Thermo Scientific™ F190
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
  • Designed for 10-32 port configurations
  • Burr and contaminant free
TRUSTED_SUSTAINABILITY
Specifications
Type High Pressure Thread Nut with Ferrule
Description Two-piece Fingerlok Nut with PEEK Ferrule
For Use With (Equipment) HPLC Injectors with 10-32 Port Configuration
Material Stainless Steel
Quantity 1 Each