missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaccinia Virus A33R Antibody [Alexa Fluor™ 532] - (Copenhagen), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43775AF532
This item is not returnable.
View return policy
Description
Vaccinia Virus A33R Polyclonal antibody specifically detects Vaccinia Virus A33R in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Vaccinia Virus A33R | |
| Polyclonal | |
| Western Blot, ELISA | |
| OPG161 | |
| Produced in rabbits immunized with purified, recombinant Vaccinia Virus A33R (Copenhagen) (Uniprot#: P68616; Val57-Asn185) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 532 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction