Learn More
UVRAG Antibody (953035) [Allophycocyanin], R&D Systems™
Product Code. 18613635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613635 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18613635 Supplier Novus Biologicals Supplier No. FAB9180A
 Restricted Item
Estimated Shipment: 09-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
UVRAG Monoclonal specifically detects UVRAG in Human samples. It is validated for CyTOF, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Antigen UVRAG
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 953035
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38
Host Species Mouse
Immunogen Human UVRAG synthetic peptide corresponding to sequences between amino acids 260-360, Accession # Q9P2Y5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cellular Signaling, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7405
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus