missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (3A8B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 533.00€
Specifications
| Antigen | Uteroglobin/SCGB1A1 |
|---|---|
| Clone | 3A8B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620266
|
Novus Biologicals
NBP2-37439-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193529
|
Novus Biologicals
NBP2-37439 |
0.1 mL |
533.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Uteroglobin/SCGB1A1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
| SCGB1A1 | |
| IgG1 | |
| Affinity Purified |
| 3A8B8 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research | |
| PBS with 0.05% Sodium Azide | |
| 7356 | |
| Purified recombinant fragment of human SCGB1A1 (AA: 26-91) expressed in E. Coli. | |
| Primary | |
| 10 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title