missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UT2/SLC14A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93774-0.02ml
This item is not returnable.
View return policy
Description
UT2/SLC14A2 Polyclonal antibody specifically detects UT2/SLC14A2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UT2/SLC14A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ16167, HUT2UT-A2, hUT-A6, MGC119566, solute carrier family 14 (urea transporter), member 2, Solute carrier family 14 member 2, urea transporter 2, Urea transporter, kidney, urea transporter-2, UT2MGC119567, UTA, UTR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 420-580 of human SLC14A2 (NP_009094.3). FRLPLSKVTYPEANRIYYLTVKSGEEEKAPSGGGGEHPPTAGPKVEEGSEAVLSKHRSVFHIEWSSIRRRSKVFGKGEHQERQNKDPFPYRYRKPTVELLDLDTMEESSEIKVETNISKTSWIRSSMAASGKRVSKALSYITGEMKECGEGLKDKSPVFQF | |
| 0.02 mL | |
| ABC Transporters, Neuroscience | |
| 8170 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction