missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP52 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94722-0.02ml
This item is not returnable.
View return policy
Description
USP52 Polyclonal antibody specifically detects USP52 in Human, Mouse samples. It is validated for Western Blot
Specifications
| USP52 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| EC 3.1.13.4, FLJ39360, hPan2, Inactive ubiquitin carboxyl-terminal hydrolase 52, KIAA0710PAB-dependent poly(A)-specific ribonuclease subunit 2, NALP4, PABP-dependent poly(A) nuclease 2, PAN2 poly(A) specific ribonuclease subunit homolog (S. cerevisiae), PAN2 polyA specific ribonuclease subunit, PYPAF4, ubiquitin specific peptidase 52, ubiquitin specific protease 52, USP52 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-1000 of human PAN2 (NP_001120932.1). DFAQVLKRSICLDQNTQAWCDTCEKYQPTIQTRNIRHLPDILVINCEVNSSKEADFWRMQAEVAFKMAVKKHGGEISKNKEFALADWKELGSPEGVLVCPSIEELKNVWLPFSIRMKMTKNKGLDVCNWTDGDEMQWGPARAEEEHGVYVYDLMATVVHILDSRTGGSLVAHIKVGETYHQRKEGVTHQQWYLFNDFLIEPIDKHEAVQFDMNWKVPAILYYVKRNLNSRYNLNIKNPIEASVLLAEASLARKQRKTHTTFIPLMLNEMPQIGDLVGLDAEFVTLNEEEAELRSDGTKSTI | |
| 0.02 mL | |
| Cell Biology | |
| 9924 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction