missing translation for 'onlineSavingsMsg'
Learn More

USP42 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18682342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18682342 0.1 mL 0.1mL
18640722 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18682342 Supplier Novus Biologicals Supplier No. NBP2943730.1ml

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP42 Polyclonal antibody specifically detects USP42 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP42
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Deubiquitinating enzyme 42, EC 3.1.2.15, EC 3.4.19.12, FLJ12697, ubiquitin carboxyl-terminal hydrolase 42, ubiquitin specific peptidase 42, ubiquitin specific protease 42, ubiquitin thioesterase 42, Ubiquitin thiolesterase 42, Ubiquitin-specific-processing protease 42
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human USP42 (NP_115548.1). ASVQNWSVNRSSVIPEHPKKQKITISIHNKLPVRQCQSQPNLHSNSLENPTKPVPSSTITNSAVQSTSNASTMSVSSKVTKPIPRSESCSQPVMNGKSKLNSSVLVPYGAESSEDSDEESKGLGKENGIGTIVSSHSPGQDAEDEEATPHELQEPMTLNGANSADSDSDPKENGLAPDGASCQGQPALHSENPFAKANGLP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84132
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.