missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP22 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94099-0.1ml
This item is not returnable.
View return policy
Description
USP22 Polyclonal antibody specifically detects USP22 in Human, Rat samples. It is validated for Western Blot
Specifications
| USP22 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human USP22 (NP_056091.1). TAEARKRKAKSCICHVCGVHLNRLHSCLYCVFFGCFTKKHIHEHAKAKRHNLAIDLMYGGIYCFLCQDYIYDKDMEIIAKEEQRKAWKMQGVGEKFSTWEP | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers | |
| 23326 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction