Learn More
USF1 Antibody (S03-5E6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19925-50ul
This item is not returnable.
View return policy
Description
USF1 Monoclonal antibody specifically detects USF1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| USF1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-5E6 | |
| Unconjugated | |
| BHLHB11, bHLHb11FCHL1, Class B basic helix-loop-helix protein 11, FCHL, HYPLIP1, Major late transcription factor 1, MLTF, MLTFI, UEF, upstream stimulatory factor 1, upstream transcription factor 1, USF | |
| A synthetic peptide of human USF1 (Uniprot # P22415) | |
| 50 μg | |
| Transcription Factors and Regulators | |
| 7391 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |