missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UROS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97299-50ul
This item is not returnable.
View return policy
Description
UROS Polyclonal antibody specifically detects UROS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UROS | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 4.2.1.75, hydroxymethylbilane hydrolyase, Hydroxymethylbilane hydrolyase [cyclizing], UROIIIS, uroporphyrinogen III synthase, Uroporphyrinogen-III cosynthase, uroporphyrinogen-III synthase | |
| Produced in rabbits immunized with E. coli-derived Human UROS fragment. | |
| 50 μg | |
| Proteases & Other Enzymes | |
| 7390 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction