missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin III Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 519.00€
Specifications
| Antigen | Uroplakin III |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673490
|
Novus Biologicals
NBP2-94533-0.02ml |
0.02 mL |
216.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685601
|
Novus Biologicals
NBP2-94533-0.1ml |
0.1 mL |
519.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uroplakin III Polyclonal antibody specifically detects Uroplakin III in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Uroplakin III | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 7380 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MGC119178, UP3a, UPIII, UPIIIA, UPK3, UPK3A, uroplakin 3, uroplakin 3A, Uroplakin III, uroplakin3a, uroplakin-3a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-207 of human UPK3A (NP_008884.1). VNLQPQLASVTFATNNPTLTTVALEKPLCMFDSKEALTGTHEVYLYVLVDSAISRNASVQDSTNTPLGSTFLQTEGGRTGPYKAVAFDLIPCSDLPSLDAIGDVSKASQILNAYLVRVGANGTCLWDPNFQGLCNAPLSAATEYRFKYVLVNMSTGLVEDQTLWSDPIRTNQLTPYSTIDTWPGRRSGG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title