missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Uroplakin II Antibody (RM507), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30733
This item is not returnable.
View return policy
Description
Uroplakin II Monoclonal antibody specifically detects Uroplakin II in Human,Macaque samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Uroplakin II | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, PBS,1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 7379 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| RM507 | |
| Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| UP2, UPII, UPK2 uroplakin 2 | |
| A peptide corresponding to residues near the C-terminus of human Uroplakin II | |
| 100 μL | |
| Primary | |
| Human, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction