missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Urocortin Monoclonal Antibody (749606)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561820
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Urocortin (UCN), a 40 amino acids neuropeptide widely distributed in the central nervous system (CNS), belongs to a family of structurally related peptides including corticotrophin releasing factor (CRF), sauvagine and urotensin I. Urocortin is derived from a larger precursor protein (pro-urocortin, also named corticotensin). Urocortin has biological actions similar to CRF such as stress-related endocrine, autonomic and behavioral responses in mammalian species. It is a potent stimulator of adrenocorticotropic hormone (ACTH) secretion by corticotroph cells from the anterior pituitary in vitro and in vivo.
Specifications
| Urocortin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P55089 | |
| Ucn | |
| E. coli-derived recombinant human Urocortin, Arg26-Lys124 (predicted). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 749606 | |
| Unconjugated | |
| Ucn | |
| Corticotensin; Mpv17; prepro-urocortin; Ucn; Ucn1; UI; UROC; Urocortin; urocortin, preproprotein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction