Learn More
Liebherr™ Upright Glass Door Lab Fridge with Interface
Product Code. 17878296
Change view
Click to view available options
Capacity (Metric):
420 L
588 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17878296 588 L Each
17868296 420 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17878296 Supplier Liebherr™ Supplier No. SRFVH5511
  • ONLINE EXCLUSIVE PRICE
    4137.00€
    3839.25€ / Pack of 1
    Save 297.75€
    7% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Secure, accurate and ultra energy-efficient fridge storage of temperature-sensitive products, the Liebherr™ Upright Glass Door Lab Fridge is fully-equipped for lab applications with Liebherr build-quality you can rely on.
  • Upright lab fridge, reach-in, 1-section
  • Temperature range 3°C to 16°C, ± 2°C temperature stability tested to EN60068-3
  • White exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration
  • Refrigerant R 600a, connection rating 2.0 A, 220-240V/50/1-ph
  • Digital controller with 2.4 in. touch and swipe color display and 0.1°C resolution, password keypad lock
  • Connectivity: Integrated LAN/WLAN interface and integrated data logger with USB-port
  • Access port for external temperature probe, additional safety thermostat
  • Alarm history with min/max temperatures, duration etc.
  • Interior lighting (refrigerator: vertical LED on left-side wall, freezer: LED on the front side at the top outside)
  • Door lock, reversible hinges, self-closing door with 90° dwell position, antimicrobial lever handle
  • Front castors with brake
  • 5-year extended warranty option available in select countries
Specifications
Type Lab Fridge
Capacity (Metric) 588 L
Temperature Range 3°C to 16°C
Temperature Sensor PT1000
Temperature (Setpoint) +5°C
Temperature Stability ±2 K
Refrigerant R 600a
Door Style Glass Self-closing
No. of Doors 1
Shelves 4
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish White Powder-coated
Interior Commercial-grade Molded Polystyrol
Certifications/Compliance ISO 9001, ISO 14001, CE
Style Upright
Display 2.4 in. Touch and Swipe Color Display in Door
Casters Front with Brake (100 mm dia.)
Color White
Condenser Type Low-maintenance Rear-wall Wire-on-tube
For Use With (Application) Storage of Temperature-sensitive Products
Ports 1 x 10 mm dia.
Product Line Scientific Perfection
Warranty 2 Years Parts and Labor, 5 Year Optional Extended Warranty
Defrost Timed Off-cycle
Model SRFvh 5511
Monitoring Options SmartCoolingHub
Smart View Compatibility Yes
Compressor CFC-free
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Wattage 120 W
Energy Usage 360 kWh/a