Learn More
UPF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23379-1-AP-150UL
 View more versions of this product
Product Code. 16852325
  • 418.21€ / 150µL
Description
Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
Specifications
Specifications
UPF1
Polyclonal
Unconjugated
UPF1
ATP dependent helicase RENT1, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1
Rabbit
Antigen Affinity Chromatography
RUO
19704, 5976, 684558
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92900, Q9EPU0
UPF1
UPF1 Fusion Protein Ag16195
150 μL
Primary
Human, Mouse, Rat
Antibody