Learn More
Kinesis™ LuerTight™ Fittings: Tefzel
Product Code. 13461299
Click to view available options
Diameter (English) Inner:
0.04 in.
0.05 in.
Color:
Black
Natural
For Use With (Equipment):
Female Luer x Female 1/4-28 Flat Bottom
To Female M6
To Male 1/4-28
Unit Size:
Each
Product Code. 13461299
Brand: Kinesis™ P624
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
TRUSTED_SUSTAINABILITY
Specifications
Type Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 80°C
Material ETFE
Color Natural
For Use With (Equipment) To Male 1/4-28