Learn More
Idex Frits: Stainless Steel
Product Code. p-68894 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
0.24 cm
Diameter (Metric) Outer:
0.64 cm
Porosity:
2 μm
Unit Size:
Pack of 10
Product Code. 11528580
Brand: Idex A100X
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Diameter (Metric) Outer 0.64 cm
Diameter (English) Inner 0.094 in.
Diameter (Metric) Inner 0.24 cm
Porosity 2 μm
Material Stainless Steel