Learn More
Kinesis™ Adapters for HPLC
Product Code. 12987739
Change view
Click to view available options
Connections:
Female Luer to Male 10-32 Adapter
Male Luer to 1/4-28 Flat Bottom
Material:
ETFE
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Material Unit Size
12987739 Female Luer to Male 10-32 Adapter ETFE Each
15136063 Male Luer to 1/4-28 Flat Bottom ETFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12987739 Supplier Kinesis™ Supplier No. 0201406
  • ONLINE EXCLUSIVE PRICE
    9.00€
    8.53€ / Pack of 1
    Save 0.47€
    5% Off
View all promotions
Estimated Shipment: 03-12-2026
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Type Adapter
Diameter (English) Inner 0.05 in.
Connections Female Luer to Male 10-32 Adapter
Material ETFE