missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Unleaded Gasoline Composite 50000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Regular Unleaded Gasoline |
| CAS | 8006-61-9 |
| Analyte or Component Names | Regular Unleaded Gasoline |
| Shipping Condition | Room Temperature |
| Concentration | 50000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Gasoline, Gasoline, natural, Natural gasoline, Petrol, natural, Gasoline, natural (10C, 11C, 14C), Oxygenate-free Rfa Gasoline, Oxygenate Free Gasoline, Unleaded, Premium |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction