Learn More
Universal Indicator, pH Range 4-10, for pH Measurement, Fisher Chemical™
Product Code. 10705351 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
Product Code. 10705351
Brand: Fisher Chemical U/0020/PB17
Request Bulk or Custom Format
Universal Indicator, pH Range 4-10, for pH Measurement
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 148741
Specifications
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Universal indicator, pH range 4-10
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol