Learn More
Fisherbrand™ Universal GC Capillary Connectors
Shop All Fisherbrand Products
Click to view available options
Connections:
Straight
Y-shaped
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Type | Universal Capillary Connector |
| Diameter (Metric) Outer | 0.30 to 0.75 mm |
| Connections | Y-shaped |
| Includes | Polypropylene box |
| Quantity | 1/Box |
| Diameter (Metric) Inner | 0.20 to 0.53 mm |
| Color | Clear |
| For Use With (Equipment) | Fused-Silica Capillary Columns in GC |