missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ULBP3 Monoclonal Antibody (166514)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560996
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
UL16-binding protein 3 (ULBP3) is an extended member of the MHC class I superfamily. It does not contain the alpha 3 domain and lacks a transmembrane domain. ULBP3 binds and activates the KLRK1/NKG2D receptor and mediates natural killer cell cytotoxicity.
Specifications
| ULBP3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BZM4 | |
| ULBP3 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-3. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 166514 | |
| Unconjugated | |
| ULBP3 | |
| ALCAN-gamma; N2DL3; N2DL-3; NKG2D ligand 3; NKG2D ligand3; NKG2DL3; RAET1N; Retinoic acid early transcript 1N; UL16 binding protein 3; UL16-binding protein 3; ULBP; ULBP 3; ULBP3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 79465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction