missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT1A4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94258-0.1ml
This item is not returnable.
View return policy
Description
UGT1A4 Polyclonal antibody specifically detects UGT1A4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UGT1A4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| bilirubin UDP-glucuronosyltransferase isozyme 2, Bilirubin-specific UDPGT isozyme 2, GNT1, hUG-BR2, polypeptide A4, UDP glucuronosyltransferase 1 family, polypeptide A4, UDP-glucuronosyltransferase 1-4, UDP-glucuronosyltransferase 1A4, UDP-glucuronosyltransferase 1-D, UDPGT, UDPGT 1-4, UGT1, UGT1*4, UGT1.4, UGT1-04, UGT-1D, UGT1DEC 2.4.1.17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-150 of human UGT1A4 (NP_009051.1). GKVLVVPTDGSPWLSMREALRELHARGHQAVVLTPEVNMHIKEEKFFTLTAYAVPWTQKEFDRVTLGYTQGFFETEHLLKRYSRSMAIMNNVSLALHRCCVELLHNEALIRHLNATSFDVVL | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 54657 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction