missing translation for 'onlineSavingsMsg'
Learn More

UFM1 Activating Enzyme/UBA5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00€ - 550.00€

Specifications

Antigen UFM1 Activating Enzyme/UBA5
Dilution Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231079
View Documents
Novus Biologicals
NBP3-35568-100ul
100 μL
550.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229083
View Documents
Novus Biologicals
NBP3-35568-20ul
20 μL
190.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

UFM1 Activating Enzyme/UBA5 Polyclonal antibody specifically detects UFM1 Activating Enzyme/UBA5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

UFM1 Activating Enzyme/UBA5
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
FLJ17281, FLJ23251UBA5, ThiFP1, UBE1DC1ubiquitin-activating enzyme E1 homolog, ubiquitin-like modifier activating enzyme 5, ubiquitin-like modifier-activating enzyme 5, UFM1-activating enzyme
Recombinant fusion protein containing a sequence corresponding to amino acids 1-404 of human UFM1 Activating Enzyme/UBA5 (NP_079094.1).,, Sequence:, MAESVERLQQRVQELERELAQERSLQVPRSGDGGGGRVRIEKMSSEVVDSNPYSRLMALKRMGIVSDYEKIRTFAVAIVGVGGVGSVTAEMLTRCGIGKLLLFDYDKVELANMNRLFFQPHQAGLSKVQAAEHTLRNINPDVLFEVHNYNITTVENFQHFMDRISNGGLEEGKPVDLVLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACFACAPPLVVAANIDEKTLKREGVCAASLPTTMGVVAGILVQNVLKFLLNFGTVSFYLGYNAMQDFFPTMSMKPNPQCDDRNCRKQQEEYKKKVAALPKQEVIQEEEEIIHEDNEWGIELVSEVSEEELKNFSGPVPDLPEGITVAYTIPKKQEDSVTELTVEDSGESLEDLMAKMKNM
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
79876
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.