missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uev1a/UBE2V1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05648-20ul
This item is not returnable.
View return policy
Description
Uev1a/UBE2V1 Polyclonal antibody specifically detects Uev1a/UBE2V1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Uev1a/UBE2V1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CROC1TRAF6-regulated IKK activator 1 beta Uev1A, CROC-1UBE2V, ubiquitin-conjugating enzyme E2 variant 1, UEV1A, UEV-1CIR1, UEV1DNA-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human Uev1a/UBE2V1 (NP_001027459.1). MAATTGSGVKVPRNFRLLEELEEGQKGVGDGTVSWGLEDDEDMTLTRWTGMIIGPPRTIYENRIYSLKIECGPKYPEAPPFVRFVTKINMNGVNSSNGVVDPRAISVLAKWQNSYSIKVVLQELRRLMMSKENMKLPQPPEGQCYSN | |
| 20 μg | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Immunology, Innate Immunity | |
| 7335 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction