missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCMA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76321-0.025mg
This item is not returnable.
View return policy
Description
UCMA Polyclonal specifically detects UCMA in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| UCMA | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C10orf49, chromosome 10 open reading frame 49, Gla-rich protein, GRP, unique cartilage matrix-associated protein, upper zone of growth plate and cartilage matrix associated | |
| Rabbit | |
| 19 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2.5-5 μg/mL, ELISA 1:100-1:2000 | |
| NP_660357 | |
| UCMA | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human UCMA. The immunogen is located within the last 50 amino acids of UCMA. Amino Acid Squence: WHYDGLHPSYLYNRHHT | |
| Affinity Purified | |
| RUO | |
| 221044 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction