missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (31A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
276.15€
Specifications
| Antigen | UCH-L1/PGP9.5 |
|---|---|
| Clone | 31A3 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Simple Western 10 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2 μg/mL, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunohistochemistry-Frozen 1-2 μg/mL |
| Applications | Western Blot |
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| UCH-L1/PGP9.5 | |
| 0.2 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| P09936 | |
| 7345 | |
| UCH-L1/PGP9.5 protein from brain (Uniprot: P09936) | |
| Primary | |
| Store at 4°C. | |
| 25 kDa |
| 31A3 | |
| Western Blot 1-2 μg/mL, Simple Western 10 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2 μg/mL, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunohistochemistry-Frozen 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
| UCHL1 | |
| IgG1 κ | |
| Protein A or G purified | |
| This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title