missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBN2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | UBN2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680932
|
Novus Biologicals
NBP2-94169-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699531
|
Novus Biologicals
NBP2-94169-0.1ml |
0.1 mL |
470.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBN2 Polyclonal antibody specifically detects UBN2 in Human samples. It is validated for Western BlotSpecifications
| UBN2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 254048 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ25778, ubinuclein 2, ubinuclein-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse UBN2 (NP_796159.3). MAEPRRVAFISLSPVRRREADFAGAEREPPRLEPQPYREPARAEPAPRADAQPPARDKPLPQREVSRAEPPMALQREPPRPEPPPPPLPLQTPPPRESAS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title