Learn More
Ubiquitin C Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-94633-0.02ml
 View more versions of this product
Product Code. 18639680
  • 201.00€ / 0.02mL
Description
Ubiquitin C Polyclonal antibody specifically detects Ubiquitin C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubiquitin C
Polyclonal
Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
HMG20, PolyUbiquitin, polyubiquitin-B, Polyubiquitin-C, UBC, Ubiquitin-40S ribosomal protein S27a, ubiquitin-60S ribosomal protein L40
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBC (NP_066289.3). MQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGGMQIFVKTLTGKTITLEVEPSDTIE
0.02 mL
Alzheimers Research, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway
7316
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat