missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin C Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94633-0.1ml
This item is not returnable.
View return policy
Description
Ubiquitin C Polyclonal antibody specifically detects Ubiquitin C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Ubiquitin C | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| HMG20, PolyUbiquitin, polyubiquitin-B, Polyubiquitin-C, UBC, Ubiquitin-40S ribosomal protein S27a, ubiquitin-60S ribosomal protein L40 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBC (NP_066289.3). MQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGGMQIFVKTLTGKTITLEVEPSDTIE | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway | |
| 7316 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction