missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 593.00€
Specifications
| Antigen | Ubiquitin |
|---|---|
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207200
|
Novus Biologicals
NBP2-54736 |
100 μL |
593.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603969
|
Novus Biologicals
NBP2-54736-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ubiquitin B Polyclonal specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Ubiquitin | |
| Polyclonal | |
| Rabbit | |
| Human | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| UBB | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 6233 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:GMQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title