missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBFD1 Polyclonal antibody specifically detects UBFD1 in Human samples. It is validated for Immunoprecipitation
Specifications
Specifications
| Antigen | UBFD1 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | FLJ42145, ubiquitin domain-containing protein UBFD1, ubiquitin family domain containing 1, Ubiquitin-binding protein homolog, UBPHFLJ38870 |
| Host Species | Rabbit |
| Immunogen | Between 259 and 309 |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?