missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98835-50ul
This item is not returnable.
View return policy
Description
UBE4A Polyclonal antibody specifically detects UBE4A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UBE4A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human UBE4A. | |
| 50 μg | |
| Cell Biology | |
| 9354 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction