Learn More
UBE3B Antibody (OTI4E4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44891-100ul
 View more versions of this product
Product Code. 30306500
  • 550.00€ / 100µL
Description
UBE3B Monoclonal antibody specifically detects UBE3B in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UBE3B
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
89910
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Western Blot
OTI4E4
Western Blot 1:2000
DKFZp586K2123, DKFZp686A1051, EC 6.3.2, EC 6.3.2.-, FLJ45294, MGC131858, MGC78388, ubiquitin protein ligase E3B, ubiquitin-protein ligase E3B
Human recombinant protein fragment corresponding to amino acids 768-1068 of human UBE3B (NP_569733) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody