missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2Q2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94350-0.1ml
This item is not returnable.
View return policy
Description
UBE2Q2 Polyclonal antibody specifically detects UBE2Q2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UBE2Q2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp762C143, EC 6.3.2.19, LOC92912, Ubiquitin carrier protein Q2, ubiquitin-conjugating enzyme E2 Q2, ubiquitin-conjugating enzyme E2Q (putative) 2, ubiquitin-conjugating enzyme E2Q family member 2, Ubiquitin-protein ligase Q2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human UBE2Q2 (NP_775740.1). MSVSGLKAELKFLASIFDKNHERFRIVSWKLDELHCQFLVPQQGSPHSLPPPLTLHCNITESYPSSSPIWFVDSEDPNLTSVLERLEDTKNNNLLRQQLKWLICELCSLYNLPKHLDVEMLDQPLPTGQNGTTEEVTSEEEEEEEEMAED | |
| 0.1 mL | |
| Cell Biology | |
| 92912 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction