missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2D4/Ubch5d Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95248-0.1ml
This item is not returnable.
View return policy
Description
UBE2D4/Ubch5d Polyclonal antibody specifically detects UBE2D4/Ubch5d in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UBE2D4/Ubch5d | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 6.3.2.19, FLJ32004, HBUCE1Ubiquitin carrier protein D4, UBCH5D, ubiquitin-conjugating enzyme E2 D4, ubiquitin-conjugating enzyme E2D 4 (putative), ubiquitin-conjugating enzyme HBUCE1, Ubiquitin-protein ligase D4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2D4 (NP_057067.1). MALKRIQKELTDLQRDPPAQCSAGPVGDDLFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTKIYHPNINSNGSICLDILRSQWSPALTVSKVLLSICSLLCDPNPDDPLVPEIAHTYKADREKYNRLAREWTQKYAM | |
| 0.1 mL | |
| Cell Biology | |
| 51619 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction