missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH10/UBE2C Mouse anti-Human, Clone: CPTC-UBE2C-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07504-100ug
This item is not returnable.
View return policy
Description
UbcH10/UBE2C Monoclonal specifically detects UbcH10/UBE2C in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications
| UbcH10/UBE2C | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C | |
| Recombinant human full-length UbcH10/UBE2C protein | |
| 100 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators | |
| 11065 | |
| Store at 4°C. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| CPTC-UBE2C-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction