missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH10/UBE2C Antibody (SAIC-41C-6) - Chimeric, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43535
This item is not returnable.
View return policy
Description
UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for Western Blot
Specifications
| UbcH10/UBE2C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot | |
| cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C | |
| Peptide LSLEFPSGYPYNAPTVK derived from the Ubiquitin-conjugating enzyme E2C, conjugated to KLH. (Uniprot# O00762) | |
| 0.2 mg | |
| Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators | |
| 11065 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| SAIC-41C-6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction