Learn More
UBC9-Specific Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67660-1-IG-150UL
This item is not returnable.
View return policy
Description
The UBC9-Specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human UBC9-Specific. This antibody recognizes human, mouse, rat antigen. The UBC9-Specific antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| UBC9-Specific | |
| Monoclonal | |
| 1900 μg/mL | |
| C358B7.1; P18; UBC9 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 18 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1G8A2 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7329 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |