missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA52 Antibody (S05-5D6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20009-100ul
This item is not returnable.
View return policy
Description
UBA52 Monoclonal antibody specifically detects UBA52 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UBA52 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-5D6 | |
| Unconjugated | |
| CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
| A synthetic peptide of human UBA52 (Uniprot # P62987) | |
| 100 μg | |
| Apoptosis, Cell Biology | |
| 7311 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction