Learn More
Ub-K63 Antibody (HWA4C4), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00199DL594
This item is not returnable.
View return policy
Description
Ub-K63 Monoclonal antibody specifically detects Ub-K63 in Human samples. It is validated for Western Blot
Specifications
| Ub-K63 | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot | |
| HWA4C4 | |
| 50mM Sodium Borate | |
| The immunogen for this antibody is Ub-K63. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |