Learn More
Ub-K63 Antibody (HWA4C4), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-00199V
Product Code. 18071219
  • 657.00€ / 100µL
Description
Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ub-K63
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
The immunogen for this antibody is Ub-K63.
100 μL
Primary
Human
Purified
Western Blot
HWA4C4
Western Blot
Mouse
Protein A or G purified
RUO
HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains.
Store at 4C in the dark.
IgG2a