missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ub-K63 Antibody (HWA4C4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00199B
This item is not returnable.
View return policy
Description
Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.
Specifications
| Ub-K63 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| The immunogen for this antibody is Ub-K63. | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HWA4C4 | |
| Western Blot | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction