missing translation for 'onlineSavingsMsg'
Learn More
Learn More
u-Plasminogen Activator/Urokinase Antibody (U-16), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
471.00€
Specifications
| Antigen | u-Plasminogen Activator/Urokinase |
|---|---|
| Clone | U-16 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 450 SE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18402936
|
Novus Biologicals
NBP1-05160MFV450 |
0.1 mL | |||||||
Description
u-Plasminogen Activator/Urokinase Monoclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
| u-Plasminogen Activator/Urokinase | |
| Western Blot | |
| mFluor Violet 450 SE | |
| Mouse | |
| Cancer | |
| 50mM Sodium Borate | |
| 5328 | |
| IgG1 | |
| Protein A or G purified |
| U-16 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator | |
| Native human uPA | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title